Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC34A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167579.1 | NP_001161051.1 | 8 | 1002 | ||
NM_003052.4 | NP_003043.3 | 8 | 1002 | |||
XM_005265975.1 | XP_005266032.1 | 7 | 945 | |||
XM_017009773.1 | XP_016865262.1 | 7 | 945 | |||
XM_017009775.1 | XP_016865264.1 | 7 | 945 | |||
GenBank mRNA | AK096088.1 | - | 8 | 981 | ||
AK298299.1 | - | 8 | 999 | |||
BC035294.1 | - | 6 | 1189 | |||
BC050385.1 | - | 8 | 979 | 5760855 | ||
BC053349.1 | - | 8 | 989 | 5186103 | ||
L13258.1 | - | 8 | 979 |