Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC18A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003054.4 | NP_003045.2 | 11, 12 | 1241 | ||
GenBank mRNA | AK314997.1 | - | 11, 12 | 1194 | ||
BC030593.1 | - | 10, 11 | 1499 | |||
BC108927.1 | - | 10, 11 | 1106 | 40025174 | ||
BC108928.2 | - | 10, 11 | 1106 | 40025175 | ||
L09118.1 | - | 11, 12 | 1183 | |||
L14269.1 | - | 11, 12 | 1111 | |||
L23205.1 | - | 11, 12 | 1191 | |||
X71354.1 | - | 11, 12 | 1141 |