Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FYTTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011537.2 | NP_001011537.2 | 4 | 487 | ||
NM_032288.6 | NP_115664.2 | 3 | 546 | |||
NR_027840.1 | 3 | 657 | ||||
GenBank mRNA | AJ344094.1 | - | 3 | 392 | ||
AK027672.1 | - | 3 | 430 | |||
AK296283.1 | - | 3 | 361 | |||
AK302546.1 | - | 3 | 657 | |||
AK303462.1 | - | 4 | 487 | |||
AK315019.1 | - | 3 | 395 | |||
AK316308.1 | - | 3 | 287 | |||
AL136558.1 | - | 3 | 399 | |||
BC035006.2 | - | 3 | 549 | 5267824 | ||
BC039734.1 | - | 3 | 456 | 6042926 | ||
CR533502.1 | - | 3 | 327 |