Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032312.3 | NP_115688.1 | 2 | 426 | ||
XM_005264599.3 | XP_005264656.1 | 2 | 420 | |||
XM_011533125.2 | XP_011531427.1 | 2 | 420 | |||
GenBank mRNA | AB072898.1 | - | 2 | 373 | ||
AK098486.1 | - | 2 | 405 | |||
BC004875.2 | - | 2 | 353 | 3687693 | ||
BP207600.1 | - | 2 | 429 |