Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032312.3 | NP_115688.1 | 6 | 1311 | ||
XM_011533125.2 | XP_011531427.1 | 5 | 1227 | |||
GenBank mRNA | AB072898.1 | - | 6 | 1258 | ||
AK098486.1 | - | 6 | 1291 | |||
BC004875.2 | - | 6 | 1238 | 3687693 |