Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032411.2 | NP_115787.1 | 2, 3 | 246 | ||
XM_006712799.3 | XP_006712862.1 | 3, 4 | 265 | |||
XM_011511993.2 | XP_011510295.1 | 2, 3 | 183 | |||
XM_017005106.1 | XP_016860595.1 | 2, 3 | 165 | |||
GenBank mRNA | AF325503.1 | - | 2, 3 | 246 | ||
AY358601.1 | - | 2, 3 | 207 | |||
BC021742.1 | - | 2, 3 | 250 | 5260075 |