Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SFT2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032740.3 | NP_116129.3 | 1 | 2330 | ||
GenBank mRNA | AK074549.1 | - | 1 | 1845 | ||
BC006808.1 | - | 1 | 687 | |||
BC063637.1 | - | 1 | 798 | |||
BC110324.1 | - | 1 | 800 |