Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318934.1 | NP_001305863.1 | 3 | 575 | ||
NM_001318935.1 | NP_001305864.1 | 3 | 575 | |||
NM_003071.3 | NP_003062.2 | 3 | 575 | |||
NM_139048.2 | NP_620636.1 | 3 | 575 | |||
XM_011513091.1 | XP_011511393.1 | 3 | 575 | |||
XM_011513092.1 | XP_011511394.1 | 3 | 575 | |||
XM_011513093.1 | XP_011511395.1 | 3 | 575 | |||
XM_017007078.1 | XP_016862567.1 | 3 | 554 | |||
XM_017007079.1 | XP_016862568.1 | 3 | 575 | |||
GenBank mRNA | AB209052.1 | - | 3 | 357 | ||
AK291231.1 | - | 3 | 522 | |||
BC005260.1 | - | 3 | 539 | |||
BC015498.1 | - | 3 | 531 | |||
BC030976.1 | - | 3 | 575 | |||
BC044659.1 | - | 3 | 543 | 6015181 | ||
L34673.1 | - | 3 | 534 | |||
S64671.1 | - | 2 | 246 |