Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052847.2 | NP_443079.1 | 5 | 959 | ||
XM_017026606.1 | XP_016882095.1 | 5 | 1068 | |||
GenBank mRNA | AB010414.1 | - | 3 | 798 | ||
BC014466.1 | - | 5 | 918 | 4861393 | ||
BC053630.1 | - | 5 | 874 | 6469872 |