Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080657.4 | NP_542388.2 | 1, 2 | 487 | ||
XM_011510414.2 | XP_011508716.1 | 2, 3 | 1576 | |||
GenBank mRNA | AF442151.1 | - | 1, 2 | 375 | ||
AK312670.1 | - | 1, 2 | 386 | |||
BC017969.1 | - | 1, 2 | 375 | 4669203 |