Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003801.1 | NP_001003801.1 | 3 | 411 | ||
NM_001003802.1 | NP_001003802.1 | 4 | 400 | |||
NM_003078.3 | NP_003069.2 | 4 | 439 | |||
XM_011516521.2 | XP_011514823.1 | 3 | 518 | |||
XM_017012556.1 | XP_016868045.1 | 3 | 238 | |||
GenBank mRNA | AK127595.1 | - | 4 | 947 | ||
AK297876.1 | - | 3 | 433 | |||
AK309500.1 | - | 3 | 396 | |||
AK316173.1 | - | 3 | 383 | |||
AY450430.1 | - | 3 | 715 | |||
AY450431.1 | - | 4 | 400 | |||
BC002628.2 | - | 4 | 400 | 3608512 | ||
BC110350.1 | - | 3 | 330 | 5212363 | ||
BI598614.1 | - | 4 | 449 | |||
BX648385.1 | - | 3 | 177 | |||
U66619.1 | - | 4 | 448 |