Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD200R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138806.3 | NP_620161.1 | 4 | 641 | ||
NM_138939.2 | NP_620385.1 | 4 | 641 | |||
NM_138940.2 | NP_620386.1 | 3 | 572 | |||
NM_170780.2 | NP_740750.1 | 3 | 572 | |||
GenBank mRNA | AF283760.1 | - | 3 | 347 | ||
AF495380.1 | - | 4 | 463 | |||
AF497548.1 | - | 3 | 394 | |||
AF497549.1 | - | 4 | 463 | |||
AF497550.1 | - | 3 | 394 | |||
AK126349.1 | - | 3 | 572 | |||
AK293071.1 | - | 4 | 586 | |||
AK304426.1 | - | 2 | 443 | |||
AY284975.1 | - | 4 | 630 | |||
AY358910.1 | - | 4 | 666 | |||
BC069661.1 | - | 4 | 439 | 7262527 | ||
BC069721.1 | - | 4 | 439 | 7262539 | ||
BC069743.1 | - | 4 | 439 | 7262551 | ||
BC093890.1 | - | 3 | 418 | 7939735 | ||
BC143393.1 | - | 4 | 487 | 9051899 |
Gene Symbol: CD200R1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008784.2 | NP_001008784.2 | 3 | 544 | ||
NM_001199215.1 | NP_001186144.1 | 4 | 613 | |||
XM_011512763.2 | XP_011511065.1 | 5 | 993 | |||
XM_017006300.1 | XP_016861789.1 | 4 | 370 | |||
GenBank mRNA | AY284976.1 | - | 4 | 613 | ||
AY552790.1 | - | 3 | 544 |