Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM175A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139076.2 | NP_620775.2 | 9 | 2692 | ||
XM_005263280.4 | XP_005263337.1 | 10 | 2825 | |||
XM_017008694.1 | XP_016864183.1 | 8 | 2598 | |||
GenBank mRNA | AK023676.1 | - | 8 | 2592 |