Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003095.2 | NP_003086.1 | 4 | 371 | ||
GenBank mRNA | AK311752.1 | - | 4 | 334 | ||
BC002505.2 | - | 4 | 364 | 3051263 | ||
BC063397.1 | - | 4 | 321 | 5176590 | ||
BC128452.1 | - | 4 | 274 | 40123237 | ||
BC128453.1 | - | 4 | 273 | 40123239 | ||
BU861031.1 | - | 4 | 374 | |||
CR456751.1 | - | 3 | 228 | |||
X85372.1 | - | 4 | 343 |