Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS2CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190707.1 | NP_001177636.1 | 22, 23 | 2663 | ||
NM_147129.4 | NP_667340.2 | 22, 23 | 2566 | |||
NR_033815.2 | 22, 23 | 2852 | ||||
NR_135622.1 | 21, 22 | 3440 | ||||
XM_005265025.1 | XP_005265082.1 | 11, 12 | 1215 | |||
GenBank mRNA | AB107015.1 | - | 22, 23 | 2554 | ||
AK074118.1 | - | 18, 19 | 3694 | |||
AK092455.1 | - | 7, 8 | 943 | |||
AK093844.1 | - | 11, 12 | 1215 | |||
AK097225.1 | - | 1, 2 | 36 | |||
AK126505.1 | - | 5, 6 | 2052 | |||
AK131270.1 | - | 22, 23 | 2660 | |||
AK131273.1 | - | 19, 20 | 2128 | |||
AL832132.1 | - | 22, 23 | 2843 | |||
AL832203.1 | - | 18, 19 | 5817 | |||
AL833107.1 | - | 1, 2 | 1399 | |||
BC042906.1 | - | 14, 15 | 1596 | |||
BC061883.1 | - | 14, 15 | 1596 | |||
BC075825.1 | - | 14, 15 | 1596 | |||
BC109233.1 | - | 6, 7 | 1018 | |||
CR627258.1 | - | 18, 19 | 5786 |