Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XXYLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308069.1 | NP_001294998.1 | 4 | 1320 | ||
NM_152531.4 | NP_689744.3 | 4 | 1783 | |||
XM_005269286.4 | XP_005269343.1 | 4 | 4168 | |||
XM_011512448.2 | XP_011510750.1 | 5 | 1595 | |||
XM_017005750.1 | XP_016861239.1 | 3 | 4556 | |||
GenBank mRNA | AK057046.1 | - | 5 | 1528 | ||
AK092474.1 | - | 2 | 1060 | |||
BC019036.2 | - | 4 | 1692 | 4591786 | ||
BC039067.1 | - | 4 | 1765 | 5185557 |