Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152544.2 | NP_689757.2 | 10 | 1957 | ||
XM_005247950.3 | XP_005248007.1 | 10 | 1954 | |||
XM_011513407.2 | XP_011511709.1 | 10 | 1986 | |||
XM_011513408.1 | XP_011511710.1 | 11 | 2098 | |||
XM_011513409.2 | XP_011511711.1 | 11 | 2069 | |||
GenBank mRNA | AK093044.1 | - | 6 | 963 |