Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039547.2 | NP_001034636.1 | 8, 9 | 906 | ||
NR_033289.1 | 8, 9 | 906 | ||||
XM_011512624.2 | XP_011510926.1 | 8, 9 | 704 | |||
XM_017006088.1 | XP_016861577.1 | 8, 9 | 891 | |||
XM_017006089.1 | XP_016861578.1 | 9, 10 | 969 | |||
GenBank mRNA | AK127641.1 | - | 8, 9 | 874 | ||
AK313792.1 | - | 8, 9 | 788 | |||
BC032470.1 | - | 8, 9 | 866 | 5217372 | ||
BX648359.1 | - | 8, 9 | 886 | |||
BX648681.1 | - | 8, 9 | 847 |