Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026832.1 | 2 | 3719 | |||
GenBank mRNA | AK055156.1 | - | 1 | 490 | ||
AK091363.1 | - | 1 | 1112 | |||
AL713643.1 | - | 1 | 1097 | |||
BC093858.1 | - | 1 | 218 |