Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276503.1 | NP_001263432.1 | 3 | 586 | ||
NM_001276504.1 | NP_001263433.1 | 3 | 614 | |||
NM_001276506.1 | NP_001263435.1 | 5 | 861 | |||
NM_003002.3 | NP_002993.1 | 4 | 731 | |||
NR_077060.1 | 5 | 785 | ||||
GenBank mRNA | AB006202.1 | - | 4 | 658 | ||
AK075360.1 | - | 4 | 657 | |||
BC005263.1 | - | 4 | 675 | 3930691 | ||
BC009574.1 | - | 4 | 656 | 4063808 | ||
BC012603.1 | - | 4 | 660 | 4108104 | ||
BC015188.1 | - | 4 | 682 | 4279214 | ||
BC015992.1 | - | 4 | 678 | 4292983 | ||
BC022350.1 | - | 4 | 670 | 4275516 | ||
BC070307.1 | - | 4 | 656 | 4277540 | ||
BC071755.1 | - | 4 | 674 | 4774525 | ||
BC071756.1 | - | 4 | 656 | 6450992 | ||
BX419391.2 | - | 4 | 709 | |||
CD105572.1 | - | 3 | 526 |