Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APCDD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304787.1 | NP_001291716.1 | 5 | 2099 | ||
NM_153360.2 | NP_699191.1 | 4 | 1928 | |||
GenBank mRNA | AK074647.1 | - | 4 | 1883 |