Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173479.3 | NP_775750.3 | 6, 7 | 896 | ||
NR_135565.1 | 6, 7 | 896 | ||||
XM_011526449.2 | XP_011524751.1 | 6, 7 | 890 | |||
GenBank mRNA | AK131444.1 | - | 6, 7 | 1255 | ||
BC031227.1 | - | 6, 7 | 896 |