Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173552.3 | NP_775823.1 | 1 | 813 | ||
XM_011512543.2 | XP_011510845.1 | 1 | 702 | |||
GenBank mRNA | AK315082.1 | - | 1 | 787 | ||
BC037293.2 | - | 1 | 540 | 5267770 |