Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UMODL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004416.2 | NP_001004416.2 | 23 | 4731 | ||
NM_001199527.1 | NP_001186456.1 | 22 | 5295 | |||
NM_001199528.2 | NP_001186457.2 | 23 | 4911 | |||
NM_173568.3 | NP_775839.3 | 22 | 5115 | |||
XM_011529797.1 | XP_011528099.1 | 20 | 4723 | |||
XM_017028506.1 | XP_016883995.1 | 22 | 5110 | |||
XM_017028507.1 | XP_016883996.1 | 22 | 8039 | |||
XM_017028508.1 | XP_016883997.1 | 12 | 2894 | |||
GenBank mRNA | AK093654.1 | - | 8 | 1973 | ||
AK124223.1 | - | 9 | 2475 | |||
AY771619.1 | - | 22 | 5115 | |||
AY771620.1 | - | 23 | 4731 |