Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001370.1 | NP_001361.1 | 9 | 1555 | ||
XM_006711956.2 | XP_006712019.1 | 9 | 1660 | |||
XM_011532649.2 | XP_011530951.1 | 9 | 1509 | |||
XM_011532650.2 | XP_011530952.1 | 9 | 1510 | |||
XM_011532652.1 | XP_011530954.1 | 9 | 1555 | |||
XM_011532653.1 | XP_011530955.1 | 9 | 1555 | |||
XM_011532654.1 | XP_011530956.1 | 8 | 1288 | |||
XM_011532655.2 | XP_011530957.1 | 5 | 577 | |||
XM_011532657.1 | XP_011530959.1 | 3 | 316 | |||
XM_011532660.1 | XP_011530962.1 | 9 | 1555 | |||
XM_017003521.1 | XP_016859010.1 | 9 | 1555 | |||
XM_017003522.1 | XP_016859011.1 | 4 | 1381 | |||
XM_017003523.1 | XP_016859012.1 | 4 | 548 | |||
XM_017003524.1 | XP_016859013.1 | 9 | 1555 | |||
GenBank mRNA | AK094676.1 | - | 5 | 768 | ||
BC104884.1 | - | 3 | 461 | |||
BC113424.1 | - | 3 | 461 | |||
BC143666.1 | - | 3 | 461 |