Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZPLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175056.1 | NP_778226.1 | 11 | 2781 | ||
XM_017005703.1 | XP_016861192.1 | 12 | 2963 | |||
XM_017005704.1 | XP_016861193.1 | 11 | 2819 | |||
GenBank mRNA | AY090780.1 | - | 11 | 2781 | ||
BC021279.2 | - | 5 | 1913 |