Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175063.5 | NP_778233.4 | 8 | 1816 | ||
NM_206538.3 | NP_996261.1 | 7 | 1729 | |||
GenBank mRNA | AL512761.1 | - | 4 | 1332 | ||
AY194293.2 | - | 12 | 2774 | |||
AY358710.1 | - | 8 | 866 | |||
AY761095.1 | - | 8 | 1794 | |||
AY761097.1 | - | 7 | 1707 | |||
BC032948.2 | - | 7 | 1668 | 4794992 | ||
BC035001.2 | - | 7 | 1666 | 4796476 | ||
BC062607.1 | - | 8 | 1958 |