Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPRIPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008949.2 | NP_001008949.1 | 3 | 418 | ||
NM_001163523.1 | NP_001156995.1 | 2 | 308 | |||
NM_001163524.1 | NP_001156996.1 | 2 | 175 | |||
NM_001324490.1 | NP_001311419.1 | 2 | 100 | |||
NM_178495.5 | NP_848590.3 | 1 | 518 | |||
XM_017003427.1 | XP_016858916.1 | 2 | 351 | |||
GenBank mRNA | BC034503.2 | - | 1 | 493 | 4824775 | |
BC073153.1 | - | 3 | 334 | 6018749 | ||
BI829328.1 | - | 1 | 518 | |||
CD300165.1 | - | 2 | 95 | |||
CN263466.1 | - | 3 | 450 |