Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182528.3 | NP_872334.2 | 2 | 1441 | ||
GenBank mRNA | AF525315.1 | - | 2 | 1440 |