Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130009.2 | NP_001123481.2 | 11 | 1348 | ||
NM_182625.4 | NP_872431.4 | 11 | 1272 | |||
XM_005262613.4 | XP_005262670.1 | 11 | 1358 | |||
XM_006712005.3 | XP_006712068.1 | 11 | 1255 | |||
XM_011532820.2 | XP_011531122.1 | 11 | 1275 | |||
XM_011532821.2 | XP_011531123.1 | 11 | 1365 | |||
XM_011532822.2 | XP_011531124.1 | 11 | 1245 | |||
GenBank mRNA | AK098188.1 | - | 4 | 298 | ||
AK131387.1 | - | 11 | 1220 | |||
BC117204.1 | - | 10 | 1170 | 40125755 | ||
BC117206.1 | - | 10 | 1170 | 40125757 | ||
CR936783.1 | - | 11 | 1358 |