Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183240.2 | NP_899063.2 | 2 | 518 | ||
XM_005263598.4 | XP_005263655.1 | 2 | 818 | |||
XM_006712300.3 | XP_006712363.1 | 2 | 642 | |||
XM_011510659.2 | XP_011508961.1 | 2 | 535 | |||
GenBank mRNA | AF361356.1 | - | 2 | 490 | ||
BC046362.1 | - | 1 | 649 | 5221396 | ||
DR003856.1 | - | 2 | 531 | |||
DR005637.1 | - | 2 | 507 |