Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194302.3 | NP_919278.2 | 23 | 3903 | ||
XM_011510903.1 | XP_011509205.1 | 23 | 3964 | |||
XM_011510904.2 | XP_011509206.1 | 23 | 5548 | |||
XM_011510908.1 | XP_011509210.1 | 23 | 3870 | |||
XM_011510909.1 | XP_011509211.1 | 23 | 3910 | |||
XM_011510910.1 | XP_011509212.1 | 23 | 3964 | |||
XM_011510911.1 | XP_011509213.1 | 23 | 3815 | |||
XM_011510912.1 | XP_011509214.1 | 23 | 3858 | |||
XM_011510914.1 | XP_011509216.1 | 22 | 3755 | |||
XM_011510915.1 | XP_011509217.1 | 22 | 3717 | |||
XM_011510916.1 | XP_011509218.1 | 22 | 3685 | |||
XM_011510919.1 | XP_011509221.1 | 23 | 3964 | |||
XM_011510921.1 | XP_011509223.1 | 13 | 2420 | |||
XM_017003752.1 | XP_016859241.1 | 24 | 4031 | |||
XM_017003753.1 | XP_016859242.1 | 23 | 5388 | |||
XM_017003754.1 | XP_016859243.1 | 22 | 3809 | |||
XM_017003755.1 | XP_016859244.1 | 22 | 5393 | |||
GenBank mRNA | AK125957.1 | - | 11 | 1692 | ||
BC130281.1 | - | 11 | 1682 |