Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198463.2 | NP_940865.1 | 7 | 624 | ||
XM_005264929.2 | XP_005264986.1 | 7 | 1004 | |||
XM_006713013.3 | XP_006713076.1 | 7 | 1004 | |||
XM_011533453.2 | XP_011531755.1 | 7 | 1004 | |||
XM_011533455.2 | XP_011531757.1 | 7 | 1004 | |||
XM_011533456.1 | XP_011531758.1 | 7 | 713 | |||
XM_011533457.1 | XP_011531759.1 | 5 | 493 | |||
XM_011533458.2 | XP_011531760.1 | 4 | 376 | |||
XM_011533459.2 | XP_011531761.1 | 7 | 1004 | |||
XM_011533460.2 | XP_011531762.1 | 2 | 143 | |||
XM_011533461.1 | XP_011531763.1 | 4 | 336 | |||
XM_017005848.1 | XP_016861337.1 | 3 | 249 | |||
XM_017005849.1 | XP_016861338.1 | 6 | 847 | |||
XM_017005850.1 | XP_016861339.1 | 6 | 525 | |||
XM_017005851.1 | XP_016861340.1 | 6 | 850 | |||
GenBank mRNA | AK124111.1 | - | 7 | 626 | ||
BC050317.1 | - | 7 | 1001 | |||
BC132815.1 | - | 5 | 438 | 40146837 | ||
BC151142.1 | - | 5 | 438 | 9020498 |