Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004699.3 | NP_004690.1 | 10, 11 | 950 | ||
GenBank mRNA | AD001530.1 | - | 10, 11 | 915 | ||
AK293119.1 | - | 10, 11 | 904 | |||
AK313697.1 | - | 10, 11 | 898 | |||
BC000028.2 | - | 10, 11 | 921 | 3505365 | ||
BC015499.2 | - | 11 | ||||
D83260.1 | - | 10, 11 | 820 |