Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003076.4 | NP_003067.3 | 6 | 870 | ||
NM_139071.2 | NP_620710.2 | 6 | 870 | |||
XM_005269107.3 | XP_005269164.2 | 5 | 747 | |||
XM_011538695.1 | XP_011536997.1 | 6 | 870 | |||
GenBank mRNA | AF109733.1 | - | 6 | 658 | ||
AK130517.1 | - | 7 | 609 | |||
AK292700.1 | - | 6 | 533 | |||
AK293932.1 | - | 6 | 728 | |||
BC009368.2 | - | 6 | 820 | 4110499 | ||
CN272379.1 | - | 5 | 400 | |||
DA825057.1 | - | 2 | 141 |