Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2AK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134335.1 | NP_001127807.1 | 5 | 680 | ||
NM_014413.3 | NP_055228.2 | 5 | 680 | |||
GenBank mRNA | AB037790.1 | - | 5 | 605 | ||
AF100784.1 | - | 5 | 569 | |||
AF147094.1 | - | 5 | 615 | |||
AF183414.1 | - | 5 | 567 | |||
AF255050.1 | - | 5 | 625 | |||
AK075192.1 | - | 5 | 608 | |||
AK290327.1 | - | 5 | 613 | |||
AK295712.1 | - | 4 | 481 | |||
AL136563.1 | - | 5 | 585 | |||
AL834494.1 | - | 5 | 585 | |||
BC006524.2 | - | 5 | 612 | 2989989 |