Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195555.1 | NP_001182484.1 | 12 | 3200 | ||
NM_001195556.1 | NP_001182485.1 | 12 | 3177 | |||
NM_014666.3 | NP_055481.1 | 12 | 3146 | |||
XM_017010087.1 | XP_016865576.1 | 12 | 3157 | |||
GenBank mRNA | AK128594.1 | - | 2 | 3428 | ||
AL833548.1 | - | 2 | 2609 | |||
BC013091.1 | - | 5 | 2012 | |||
BC030264.1 | - | 4 | 3102 | |||
D79993.1 | - | 12 | 3042 |