Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KIAA0141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014773.4 | NP_055588.3 | 12 | 1728 | ||
XM_005268552.2 | XP_005268609.1 | 11 | 1603 | |||
XM_006714812.3 | XP_006714875.1 | 6 | 1069 | |||
GenBank mRNA | AK292869.1 | - | 12 | 1677 | ||
BC007855.2 | - | 12 | 1704 | 4298364 | ||
BC010452.1 | - | 12 | 1619 | 4155782 | ||
BC011269.1 | - | 12 | 1619 | 4155755 | ||
BC011926.1 | - | 12 | 1649 | 4543650 | ||
BC035486.1 | - | 12 | 1644 | |||
D50931.1 | - | 12 | 1686 |