Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161661.1 | NP_001155133.1 | 3 | 416 | ||
NM_001161662.1 | NP_001155134.1 | 3 | 416 | |||
NM_015238.2 | NP_056053.1 | 3 | 416 | |||
XM_005265850.1 | XP_005265907.1 | 3 | 373 | |||
XM_005265853.2 | XP_005265910.1 | 3 | 373 | |||
XM_011534485.1 | XP_011532787.1 | 3 | 373 | |||
XM_011534486.1 | XP_011532788.1 | 3 | 373 | |||
XM_011534487.1 | XP_011532789.1 | 3 | 373 | |||
XM_011534488.1 | XP_011532790.1 | 3 | 373 | |||
XM_011534489.1 | XP_011532791.1 | 3 | 373 | |||
XM_011534490.1 | XP_011532792.1 | 3 | 373 | |||
XM_011534491.1 | XP_011532793.1 | 3 | 373 | |||
XM_017009276.1 | XP_016864765.1 | 3 | 373 | |||
XM_017009277.1 | XP_016864766.1 | 3 | 736 | |||
XM_017009278.1 | XP_016864767.1 | 3 | 808 | |||
GenBank mRNA | AF506799.1 | - | 3 | 389 | ||
AK126376.1 | - | 3 | 389 | |||
AK296323.1 | - | 3 | 326 | |||
BX640827.1 | - | 3 | 220 |