Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016048.2 | NP_057132.2 | 5 | 994 | ||
GenBank mRNA | AF151869.1 | - | 5 | 998 | ||
AK025787.1 | - | 5 | 992 | |||
BC008367.1 | - | 5 | 972 | 3355904 | ||
BC014105.2 | - | 5 | 982 | 4129769 |