Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016484.4 | NP_057568.1 | 5, 6 | 433 | ||
XM_011530973.2 | XP_011529275.1 | 6, 7 | 597 | |||
XM_017029568.1 | XP_016885057.1 | 5, 6 | 764 | |||
GenBank mRNA | AF151061.1 | - | 5, 6 | 403 | ||
AK024746.1 | - | 5, 6 | 401 | |||
AK311539.1 | - | 4, 5 | 295 | |||
AY358829.1 | - | 5, 6 | 639 | |||
BC012996.2 | - | 5, 6 | 372 | 3530647 | ||
BC089433.1 | - | 4, 5 | 330 | |||
BG708698.1 | - | 5, 6 | 437 | |||
BX537725.1 | - | 5, 6 | 507 | |||
CR457149.1 | - | 4, 5 | 331 |