Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160354.1 | NP_001153826.1 | 2 | 617 | ||
NM_001160355.1 | NP_001153827.1 | 2 | 617 | |||
NM_017527.3 | NP_059997.3 | 2 | 617 | |||
XM_005250989.4 | XP_005251046.1 | 2 | 805 | |||
XM_005250990.4 | XP_005251047.1 | 2 | 937 | |||
GenBank mRNA | AB105187.1 | - | 2 | 808 | ||
AI740834.1 | - | 2 | 370 | |||
AJ001348.1 | - | 2 | 629 | |||
AK092545.1 | - | 2 | 256 | |||
BC001291.1 | - | 2 | 243 | |||
BC117142.1 | - | 2 | 430 | 40125693 | ||
BC117144.1 | - | 2 | 430 | 40125695 | ||
BG491223.1 | - | 2 | 617 |