Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034915.2 | NP_001030087.2 | 16 | 2429 | ||
NM_001122825.1 | NP_001116297.1 | 14 | 2180 | |||
NM_001122826.1 | NP_001116298.1 | 15 | 2319 | |||
NM_001122827.1 | NP_001116299.1 | 14 | 2168 | |||
NM_017697.3 | NP_060167.2 | 16 | 2441 | |||
XM_005250991.3 | XP_005251048.1 | 15 | 2468 | |||
XM_005250992.3 | XP_005251049.1 | 15 | 2427 | |||
GenBank mRNA | AK000178.1 | - | 12 | 1726 | ||
AK001805.1 | - | 3 | 467 | |||
BC019932.2 | - | 14 | 2141 | |||
BC035406.1 | - | 12 | 1703 | |||
BC067098.1 | - | 16 | 2395 | 6586195 | ||
BC099916.1 | - | 16 | 2393 | 30527936 | ||
BG182742.1 | - | 1 | 180 | |||
BG212336.1 | - | 1 | 180 | |||
BX641160.1 | - | 1 | 443 | |||
BX647570.1 | - | 15 | 2381 |