Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018227.5 | NP_060697.4 | 9 | 823 | ||
XM_017008359.1 | XP_016863848.1 | 9 | 800 | |||
GenBank mRNA | AB014773.1 | - | 9 | 771 | ||
AK094164.1 | - | 1 | 29 | |||
AK094969.1 | - | 9 | 800 | |||
AK314371.1 | - | 9 | 800 | |||
AL832015.2 | - | 9 | 792 | |||
AL832458.2 | - | 9 | 792 | |||
BC031637.1 | - | 9 | 805 | |||
BC126484.1 | - | 9 | 818 | 8992200 | ||
BC126486.1 | - | 9 | 818 | 8992202 | ||
EF623993.1 | - | 9 | 771 |