Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018238.3 | NP_060708.1 | 2 | 255 | ||
XM_005250023.3 | XP_005250080.1 | 2 | 151 | |||
XM_011516397.2 | XP_011514699.1 | 2 | 721 | |||
GenBank mRNA | AJ278150.1 | - | 1 | 111 | ||
AK001704.1 | - | 2 | 123 | |||
AK222769.1 | - | 2 | 117 | |||
AK299131.1 | - | 2 | 123 | |||
AK304502.1 | - | 2 | 138 | |||
BC009775.1 | - | 1 | 115 | |||
BC022777.1 | - | 1 | 109 | 3913448 | ||
DB032353.1 | - | 2 | 255 |