Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171080.1 | NP_001164551.1 | 5 | 757 | ||
NM_001324242.1 | NP_001311171.1 | 6 | 829 | |||
NM_001324243.1 | NP_001311172.1 | 6 | 922 | |||
NM_001324244.1 | NP_001311173.1 | 6 | 829 | |||
NM_018301.4 | NP_060771.3 | 5 | 757 | |||
XM_011530980.2 | XP_011529282.1 | 6 | 921 | |||
XM_011530981.2 | XP_011529283.1 | 5 | 849 | |||
GenBank mRNA | AK001878.1 | - | 5 | 730 | ||
BC006986.1 | - | 5 | 734 | 3995903 |