Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018475.4 | NP_060945.2 | 6 | 1448 | ||
NR_073070.1 | 7 | 1551 | ||||
XM_017008412.1 | XP_016863901.1 | 8 | 1366 | |||
GenBank mRNA | AF132746.1 | - | 6 | 1171 | ||
AF183409.1 | - | 6 | 1171 | |||
AK026163.1 | - | 5 | 831 | |||
AK027162.1 | - | 1 | 2112 | |||
AK290663.1 | - | 6 | 1401 | |||
AK295289.1 | - | 7 | 1501 | |||
BC104978.1 | - | 6 | 1244 | 8143981 | ||
BC104980.1 | - | 6 | 1246 | 8143983 | ||
BC107582.1 | - | 6 | 1408 | 4691718 |