Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAGE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018666.2 | NP_061136.2 | 14 | 1814 | ||
XM_017029621.1 | XP_016885110.1 | 14 | 1822 | |||
XM_017029622.1 | XP_016885111.1 | 14 | 1733 | |||
XM_017029623.1 | XP_016885112.1 | 14 | 1696 | |||
XM_017029624.1 | XP_016885113.1 | 13 | 1636 | |||
XM_017029625.1 | XP_016885114.1 | 14 | 1729 | |||
XM_017029626.1 | XP_016885115.1 | 8 | 938 | |||
XM_017029627.1 | XP_016885116.1 | 4 | 729 | |||
GenBank mRNA | AJ278111.1 | - | 14 | 1814 | ||
AK310412.1 | - | 13 | 1770 | |||
BC144260.1 | - | 7 | 567 | 9052780 | ||
CD358947.1 | - | 6 | 652 |