Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018843.3 | NP_061331.2 | 7 | 805 | ||
GenBank mRNA | AF125531.1 | - | 8 | 778 | ||
AK223020.1 | - | 7 | 759 | |||
AK290848.1 | - | 7 | 580 | |||
BC027322.1 | - | 6 | 537 | 4345193 |