Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018843.3 | NP_061331.2 | 12 | 1391 | ||
GenBank mRNA | AK223020.1 | - | 12 | 1345 | ||
AK290848.1 | - | 12 | 1166 | |||
BC027322.1 | - | 11 | 1123 | 4345193 |